4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester 95%

Supplier: Thermo Scientific

H27097.MD H27097.03
H27097.MDEA 76.3 GBP
H27097.MD H27097.03
4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester 95%
4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester

Purity: 95% CAS No.: 165881-36-7 Molecular Formula: C14H24BBrO2 Formula Weight: 315.06 Refractive Index: 1.4960 Optical Rotation: +18° (c=1 in chloroform) MDL No.: MFCD09037500

Formula: C₁₄H₂₄BBrO₂
MW: 315,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037500
CAS Number: 165881-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR