2-Phenylthioacetamide 97%

Supplier: Thermo Fisher Scientific
Danger

H26514.MD H26514.03
H26514.MDEA 25.7 GBP
H26514.MD H26514.03
2-Phenylthioacetamide 97%
2-Phenylthioacetamide

Purity: 97% CAS No.: 645-54-5 Molecular Formula: C8H9NS Formula Weight: 151.23 Melting Point: 91-95° MDL No.: MFCD00022177

Formula: C₈H₉NS
Melting Pt: 91…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00022177
CAS Number: 645-54-5
EINECS: None

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR