tert-Butyl-N-allylcarbamate 97%

Supplier: Thermo Scientific

Synonyms: N-Boc-allylamine, N-(tert-Butoxycarbonyl)allylamine, N-Allylcarbamic Acid tert-Butyl Ester, tert-Butyl allylcarbamate

H26014.09 H26014.18
H26014.09EA 60.3 GBP
H26014.09 H26014.18
tert-Butyl-N-allylcarbamate 97%
tert-Butyl-N-allylcarbamate

Purity: 97% CAS No.: 78888-18-3 Molecular Formula: C8H15NO2 Formula Weight: 157.21 Flash Point: 79°(175°F) Melting Point: 34-39° Boiling Point: 48-50°/1mm MDL No.: MFCD00191870

Formula: C₈H₁₅NO₂
MW: 157,21 g/mol
Boiling Pt: 48…50 °C (1 mmHg)
Melting Pt: 34…39 °C
Density: 0,938
Flash Pt: 79 °C (175 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191870
CAS Number: 78888-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR