2-Furamide 97%

Supplier: Thermo Scientific

Synonyms: Furan-2-carboxamide

B25075.03 B25075.06 B25075.14
B25075.03EA 24.6 GBP
B25075.03 B25075.06 B25075.14
2-Furamide 97%
2-Furamide

Purity: 97% CAS No.: 609-38-1 Molecular Formula: C5H5NO2 Formula Weight: 111.10 Melting Point: 139-142° MDL No.: MFCD00236147

Formula: C₅H₅NO₂
MW: 111,1 g/mol
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00236147
CAS Number: 609-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR