Ergosterol hydrate, (max. 6% H₂O) 96% (dry weight)
Supplier: Thermo Fisher Scientific
|
Danger
|
B23840.14
B23840.06
B23840.14EA
143
GBP
B23840.14
B23840.06
Ergosterol hydrate, (max. 6% H₂O) 96% (dry weight)
Ergosterol hydrate
Purity: 96%
CAS No.: 57-87-4
Molecular Formula: C28H44O.xH2O
Formula Weight: 396.65(anhy)
Storage Temperature: 2°C to 8°C
Melting Point: 154-160°
UN No.: UN2811
MDL No.: MFCD00003623
Applications: A membrane lipid found almost exclusively in fungi. It is used as an indicator of living fungal biomass
Formula:
C₂₈H₄₄O·xH₂O Melting Pt: 154…160 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003623 CAS Number: 57-87-4 EINECS: 200-352-7 UN: 2811 ADR: 6.1,II Merck Index: 14,3659 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...