N-Benzylformamide 99%

Supplier: Thermo Fisher Scientific

B23722.06 B23722.14 B23722.22
B23722.06EA 43.2 GBP
B23722.06 B23722.14 B23722.22
N-Benzylformamide 99%
N-Benzylformamide

Purity: 99% CAS No.: 6343-54-0 Molecular Formula: C6H5CH2NHOCH Formula Weight: 135.17 Melting Point: 60-61° MDL No.: MFCD00003281

Formula: C₈H₉NO
Melting Pt: 60…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00003281
CAS Number: 6343-54-0
EINECS: 228-739-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR