Benzyl-6-hydroxynicotinate 99%
Supplier: Thermo Scientific
|
Warning
|
B21807.14
B21807.06
B21807.14EA
118
GBP
B21807.14
B21807.06
Benzyl-6-hydroxynicotinate 99%
Benzyl-6-hydroxynicotinate
Purity: 99%
CAS No.: 191157-01-4
Molecular Formula: C13H11NO3
Formula Weight: 229.24
Melting Point: 178-181°
MDL No.: MFCD02683072
Formula:
C₁₃H₁₁NO₃ MW: 229,24 g/mol Melting Pt: 178…181 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683072 CAS Number: 191157-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...