Ethyl-3-(2-furyl)propionate 98%
Supplier: Thermo Scientific
A18732.14
A18732.06
A18732.14EA
103
GBP
A18732.14
A18732.06
Ethyl-3-(2-furyl)propionate 98%
Ethyl-3-(2-furyl)propionate
Purity: 98%
CAS No.: 10031-90-0
Molecular Formula: C9H12O3
Formula Weight: 168.19
MDL No.: MFCD00036496
Formula:
C₉H₁₂O₃ MW: 168,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00036496 CAS Number: 10031-90-0 EINECS: 233-097-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...