2-Phenylbutyronitrile 95%

Supplier: Thermo Fisher Scientific
Danger

A17093.09 A17093.18
A17093.09EA 25.8 GBP
A17093.09 A17093.18
2-Phenylbutyronitrile 95%
2-Phenylbutyronitrile

Purity: 95% CAS No.: 769-68-6 Molecular Formula: C10H11N Formula Weight: 145.20 Flash Point: 105°(221°F) Refractive Index: 1.5086 Boiling Point: 102-104°/7mm UN No.: UN3276 MDL No.: MFCD00001876

Formula: C₁₀H₁₁N
Boiling Pt: 102…104 °C (7 mmHg)
Density: 0,975
Flash Pt: 105 °C (221 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001876
CAS Number: 769-68-6
EINECS: 212-213-8
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR