5-Acetylvaleric acid 96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
6-Oxoheptanoic acid
A14810.06
A14810.09
A14810.18
A14810.06EA
55.7
GBP
A14810.06
A14810.09
A14810.18
5-Acetylvaleric acid 96%
5-Acetylvaleric acid
Purity: 96%
CAS No.: 3128-07-2
Molecular Formula: C7H12O3
Formula Weight: 144.17
Refractive Index: 1.4490
Melting Point: 34-36°
Boiling Point: 138-140°/1.5mm
Solubility: Soluble in water
Beilstein Reference: 1362931
UN No.: UN3261
MDL No.: MFCD00051479
Formula:
C₇H₁₂O₃ MW: 144,17 g/mol Boiling Pt: 138…140 °C (1,5 mmHg) Melting Pt: 34…36 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051479 CAS Number: 3128-07-2 EINECS: 221-512-2 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...