Crocein scarlet 3B

Supplier: Thermo Fisher Scientific

Synonyms: Acid Red 73

A14642.18 A14642.30
A14642.18EA 30.4 GBP
A14642.18 A14642.30
Crocein scarlet 3B
Crocein scarlet 3B

CAS No.: 5413-75-2 Molecular Formula: C22H14N4Na2O7S2 Formula Weight: 556.49 Solubility: Soluble in water MDL No.: MFCD00003907

Formula: C₂₂H₁₆N₄O₇S₂
MW: 556.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003907
CAS Number: 5413-75-2
EINECS: 226-502-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR