3-Amino-4-methoxybenzamide 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Amino-p-anisamide
A13942.06
A13942.14
A13942.06EA
35
GBP
A13942.06
A13942.14
3-Amino-4-methoxybenzamide 98%
3-Amino-4-methoxybenzamide
Purity: 98%
CAS No.: 17481-27-5
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Melting Point: 156-160°
MDL No.: MFCD00017132
Formula:
C₈H₁₀N₂O₂ MW: 166,18 g/mol Melting Pt: 156…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017132 CAS Number: 17481-27-5 EINECS: 241-497-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...