Acetone semicarbazone ≥96%

Supplier: Thermo Scientific
Danger

Synonyms: 2-(Propan-2-ylidene)hydrazinecarboxamide

A12436.30 A12436.36
A12436.30EA 0 GBP
A12436.30 A12436.36
Acetone semicarbazone ≥96%
Acetone semicarbazone

Purity: 96% CAS No.: 110-20-3 Molecular Formula: C4H9N3O Formula Weight: 115.14 Melting Point: 179-185° Beilstein Reference: 970110 MDL No.: MFCD00014785

Formula: C₄H₉N₃O
MW: 115,14 g/mol
Melting Pt: 179…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00014785
CAS Number: 110-20-3
EINECS: 203-746-7
Merck Index: 14,8444

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR