Phenylfluorone

Supplier: Thermo Scientific

Synonyms: 9-Phenyl-2,3,7-trihydroxy-6-fluorone, 9-Phenyl-3-fluorone, 2,6,7-Trihydroxy-9-phenyl-3-isoxanthone

A11495.06 A11495.14 A11495.22
A11495.06EA 50.2 GBP
A11495.06 A11495.14 A11495.22
Phenylfluorone
Phenylfluorone

CAS No.: 975-17-7 Molecular Formula: C19H12O5 Formula Weight: 320.30 Melting Point: 300° Beilstein Reference: 40910 MDL No.: MFCD00005048

Formula: C₁₉H₁₂O₅
MW: 320,30 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005048
CAS Number: 975-17-7
EINECS: 213-550-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR