4-Methylbenzoylacetonitrile 97%

Supplier: Thermo Scientific
Warning

A10950.03 A10950.06 A10950.14
A10950.03EA 31.8 GBP
A10950.03 A10950.06 A10950.14
4-Methylbenzoylacetonitrile 97%
4-Methylbenzoylacetonitrile

Purity: 97% CAS No.: 7391-28-8 Molecular Formula: C10H9NO Formula Weight: 159.19 Melting Point: 99-104° Beilstein Reference: 742923 MDL No.: MFCD00067895

Formula: C₁₀H₉NO
MW: 159,188 g/mol
Melting Pt: 99…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00067895
CAS Number: 7391-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR