QuadraSil™ SA metal scavenger

Supplier: Thermo Scientific

47264.14
47264.14EA 0 GBP
47264.14
QuadraSil™ SA metal scavenger
QuadraSil™ SA

Physical Form: Beads UN No.:UN3077

UN: 3077
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR