Manganesebis(trifluoromethylsulphonyl)imide

Supplier: Thermo Scientific
Danger

Synonyms: Manganese(II) triflimide, Manganese(II) Bis(trifluoromethanesulphonyl)imide, Manganese bis(trifluoromethanesulphonyl)imide, Bis(trifluoromethanesulphonyl)imide manganese(II) salt

47179.MD 47179.03 47179.06
47179.MDEA 0 GBP
47179.MD 47179.03 47179.06
Manganesebis(trifluoromethylsulphonyl)imide
Manganesebis(trifluoromethylsulphonyl)imide

CAS No.: 207861-55-0 Molecular Formula: C4F12MnN2O8S4 Formula Weight: 615.21 Physical Form: Solid Sensitivity: Hygroscopic Appearance: Off-white UN No.: UN3261 MDL No.: MFCD23380173

Formula: C₄F₁₂MnN₂O₈S₄
MW: 615,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23380173
CAS Number: 207861-55-0
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR