Smopex®-102 metal scavenger

Supplier: Thermo Scientific

44705.22EA 0 GBP
44705.22
Smopex®-102 metal scavenger
Smopex®-102

Applications: Recovery of precious metals from catalyst process streams, especially Pt. Also, base metals removed: Fe, Cu, and Ni

Density: 0,3 g/mL
MDL Number: MFCD06202259
CAS Number: 289911-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR