Smopex®-101 metal scavenger
Supplier: Thermo Scientific
44568.22
44568.22EA
0
GBP
44568.22
Smopex®-101 metal scavenger
Smopex®-101
CAS No.: 194498-37-8
Physical Form: Fibers
Appearance: Yellow to brown
Odour: Odourless
Melting Point: 120°
MDL No.: MFCD06202258
Applications: Recovery of precious metals from catalyst process streams, especially Pd, Rh, Ru, Pt, and Ir. Also, base metals removed: Ag, Fe, Cu, Al, Ni, and V.
Melting Pt:
120 °C |
MDL Number:
MFCD06202258 CAS Number: 194498-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...