Tetraammineplatinum(II) hydrogen phosphate solution (0,5% Pt) (w/w)
Supplier: Thermo Scientific
|
Danger
|
44076.NB
44076.NH
44076.NBEA
720
GBP
44076.NB
44076.NH
Tetraammineplatinum(II) hydrogen phosphate solution (0,5% Pt) (w/w)
Tetraammineplatinum(II) hydrogen phosphate
CAS No.: 127733-98-6
Molecular Formula: H13N4O4PPt
Formula Weight: 359.18
Physical Form: Liquid
Density: 1.013
Appearance: Colourless
MDL No.: MFCD03788258
Formula:
H₁₃N₄O₄PPt MW: 359,18 g/mol Density: 1,013 Storage Temperature: Ambient |
MDL Number:
MFCD03788258 CAS Number: 127733-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...