B-Tribromoborazine ≥96%
Supplier: Thermo Fisher Scientific
|
Danger
|
43723.03
43723.06
43723.03EA
82.6
GBP
43723.03
43723.06
B-Tribromoborazine ≥96%
B-Tribromoborazine
Purity: 96%
CAS No.: 13703-88-3
Molecular Formula: H3B3Br3N3
Formula Weight: 317.20
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 124-128°
UN No.: UN1759
MDL No.: MFCD00058924
Formula:
B₃Br₃H₃N₃ MW: 317.187 g/mol Melting Pt: 124…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00058924 CAS Number: 13703-88-3 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...