Methyl-3-oxotetrahydrothiophene-2-carboxylate ≥98%

Supplier: Thermo Fisher Scientific
Warning

43634.06 43634.03 43634.MD
43634.06EA 0 GBP
43634.06 43634.03 43634.MD
Methyl-3-oxotetrahydrothiophene-2-carboxylate ≥98%
Methyl-3-oxotetrahydrothiophene-2-carboxylate

Purity: 98% CAS No.: 2689-69-2 Molecular Formula: C6H8O3S Formula Weight: 160.19 Physical Form: Liquid MDL No.: MFCD00156156

Formula: C₆H₈O₃S
Storage Temperature: Ambient
MDL Number: MFCD00156156
CAS Number: 2689-69-2
EINECS: 220-257-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR