Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 14,000
Supplier: Thermo Scientific
|
Warning
|
43230.22
43230.14
43230.06
43230.22EA
218
GBP
43230.22
43230.14
43230.06
Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 14,000
Poly(methyl-3,3,3-trifluoropropylsiloxane)
CAS No.: 63148-56-1
Molecular Formula: [-Si(CH3)(CH3CH2CF3)O-]n
Flash Point: 315°(599°F)
Physical Form: Liquid
Density: 1.30
Refractive Index: 1.383
MDL No.: MFCD00241477
Applications: Fluorosilicone lubricant
Formula:
[-Si(CH₃)(CH₃CH₂CF₃)O-]n Density: 1,30 Flash Pt: 315 °C (599 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00241477 CAS Number: 63148-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...