Aluminium oxide 10% (w/v) in toluene polymeric precursor

Supplier: Thermo Scientific
Danger

Synonyms: Aluminium(III) oxide, Alumina C

42767.AK 42767.AD
42767.AKEA 676 GBP
42767.AK 42767.AD
Aluminium oxide 10% (w/v) in toluene polymeric precursor
Aluminium oxide

Physical Form: Viscous liquid/semi-solid Sensitivity: Moisture Sensitive Solubility: Soluble in hexane, toluene UN No.: UN1993 MDL No.: MFCD00003424


Applications: Aluminum oxide sealants/films

Formula: Al₂O₃
MW: 101,96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR