Analcime naturally occurring mineral, grains ∼ 0.06-0.19 in

Supplier: Thermo Fisher Scientific

42524.06 42524.14
42524.06EA 0 GBP
42524.06 42524.14
Analcime naturally occurring mineral, grains ∼ 0.06-0.19 in
Analcime

Molecular Formula: Na2O.Al2O3.4SiO2.2H2O Formula Weight: 476.28 (440.28anhy) MDL No.: MFCD01864014


Applications: For grain mounts, XRD and microprobe standards, indentification of unknown minerals

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR