Inhibitor removal resin
Supplier: Thermo Fisher Scientific
42489.22
42489.36
42489.14
42489.22EA
73.4
GBP
42489.22
42489.36
42489.14
Inhibitor removal resin
Inhibitor removal resin
Applications: For the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol
Storage Temperature:
Ambient |
MDL Number:
MFCD00081548 CAS Number: 9003-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...