Chloropentaamminerhodium(III) chloride (≥34,5% Rh)
Supplier: Thermo Scientific
|
Warning
|
10519.02
10519.03
10519.06
10519.02EA
401
GBP
10519.02
10519.03
10519.06
Chloropentaamminerhodium(III) chloride (≥34,5% Rh)
Chloropentaamminerhodium(III) chloride
CAS No.: 13820-95-6
Molecular Formula: [RhCl(NH3)5]Cl2
Formula Weight: 294.42
Physical Form: Micro Crystals
Sensitivity: Hygroscopic
Melting Point: dec.
MDL No.: MFCD00135665
Formula:
[RhCl(NH₃)₅]Cl₂ Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00135665 CAS Number: 13820-95-6 EINECS: 237-505-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...