Anti-KIT Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP3797AEA 339 GBP
ABGEAP3797A
Anti-KIT Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KIT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR