VWR®, Marker Pen
Supplier: VWR
VWR®
631-1136EA 20.7 GBP
631-1136
VWR®, Marker Pen
Pens Permanent Markers
The ultra-fine tip of this permanent marker, and the contrasting black colour, allow clearly legible writing. Writes on glass, plastic and paper.
  • Writes immediately, no priming required
  • Solvent resistant
  • Odourless
  • Fast drying
  • Holding clip to attach it to a pocket or writing board