Safety shoes, sandals, Velcro® fastening, Style & Job, alligator
Supplier: U-Power
BC30355/35 BC30355/36 BC30355/37 BC30355/38 BC30355/39 BC30355/40 BC30355/41 BC30355/42 BC30355/43 BC30355/44 BC30355/45 BC30355/46 BC30355/47 BC30355/48
UPOWBC30355/35EA 54.9 GBP
UPOWBC30355/35 UPOWBC30355/36 UPOWBC30355/37 UPOWBC30355/38 UPOWBC30355/39 UPOWBC30355/40 UPOWBC30355/41 UPOWBC30355/42 UPOWBC30355/43 UPOWBC30355/44 UPOWBC30355/45 UPOWBC30355/46 UPOWBC30355/47 UPOWBC30355/48
Safety shoes, sandals, Velcro® fastening, Style & Job, alligator
Shoes and Boots
Anti-static safety shoes made from water resistant leather and a slip-resistant and hard wearing PU sole.
  • Removable anatomical FlatFit insole
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Certifications: EN 20345:2011; S1P, SRC