Adapters, glass, expansion
Supplier: SMITH SCIENTIFIC
8SXAE10 8SXAE21 8SXAE31 8SXAE32 8SXAE41 8SXAE42 8SXAE43 8SXAE52 8SXAE53 8SXAE54
SMIT8SXAE10EA 69.6 GBP
SMIT8SXAE10 SMIT8SXAE21 SMIT8SXAE31 SMIT8SXAE32 SMIT8SXAE41 SMIT8SXAE42 SMIT8SXAE43 SMIT8SXAE52 SMIT8SXAE53 SMIT8SXAE54
Adapters, glass, expansion
Adapters Adapters, Conical Ground Joints
Borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.