Anti-FTO Mouse Monoclonal Antibody [clone: FT 62-6]

Supplier: Cayman Chemical
10814-50 10814-100
CAYM10814-50EA 542 GBP
CAYM10814-50 CAYM10814-100
Anti-FTO Mouse Monoclonal Antibody [clone: FT 62-6]
Antibodies
Type: Primary
Antigen: FTO
Clonality: Monoclonal
Clone: FT 62-6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR