Anti-His-Tag Mouse Monoclonal Antibody [clone: 12K267]

Supplier: United States Biological
USBIH5099-05D-100EA 412 GBP
USBIH5099-05D-100
Anti-His-Tag Mouse Monoclonal Antibody [clone: 12K267]
Antibodies
Type: Tag
Antigen: His-Tag
Clonality: Monoclonal
Clone: 12K267
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR