Anti-CD90.2 Rat Monoclonal Antibody [clone: 30-H12]

Supplier: US Biological
C2441-05C-500 C2441-05C-50
USBIC2441-05C-500EA 480 GBP
USBIC2441-05C-500 USBIC2441-05C-50
Anti-CD90.2 Rat Monoclonal Antibody [clone: 30-H12]
Antibodies
Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 30-H12
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR