Anti-Heat Shock Protein 27, Rabbit Polyclonal Antibody

Supplier: US Biological
H1830-52L-100 H1830-52L-25
USBIH1830-52L-100EA 289 GBP
USBIH1830-52L-100 USBIH1830-52L-25
Anti-Heat Shock Protein 27, Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:2000; Immunoprecipitation: 1:100

Type: Primary
Antigen: Heat Shock Protein 27,
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Guinea pig, Hamster, Human, Monkey, Pig, Rat, Sheep,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR