Anti-IgG (H+L) Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Agrisera
AGRIAS10833EA 212 GBP
AGRIAS10833
Anti-IgG (H+L) Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR