Biopsy inserts, stainless steel
Supplier: Epredia
720-0369EA 0 GBP
720-0369
Biopsy inserts, stainless steel
Embedding Cassettes Inserts for Biopsy / Embedding Cassettes
Biopsy inserts are two-part loose fitting stainless steel capsules with welded mesh.
  • Mesh fits inside processing cassettes
  • Prevent the loss of materials
Mesh hole size is 0,5 mm