Anti-IL21 Rabbit Polyclonal Antibody

Supplier: US Biological
I8442-05
USBII8442-05EA 458 GBP
USBII8442-05
Anti-IL21 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:50000; Immunochemistry: 1:500

Type: Primary
Antigen: IL21 (interleukin 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR