Anti-CD47 Mouse Monoclonal Antibody [clone: 8J217]

Supplier: United States Biological
USBIC2402-03FEA 601 GBP
USBIC2402-03F
Anti-CD47 Mouse Monoclonal Antibody [clone: 8J217]
Antibodies
Type: Primary
Antigen: CD47 (CD47 molecule)
Clonality: Monoclonal
Clone: 8J217
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR