Anti-Rift Valley Fever Virus Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI146247EA 493 GBP
USBI146247
Anti-Rift Valley Fever Virus Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1 ?g/ml

Type: Primary
Antigen: Rift Valley Fever Virus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR