Anti-MT2A Mouse Monoclonal Antibody [clone: 6G2]

Supplier: US Biological
129947
USBI129947EA 489 GBP
USBI129947
Anti-MT2A Mouse Monoclonal Antibody [clone: 6G2]
Antibodies
Type: Primary
Antigen: MT2A (metallothionein 2A)
Clonality: monoclonal
Clone: 6G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Lambda
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR