Anti-ATP1B2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
032271-APC
USBI032271-APCEA 798 GBP
USBI032271-APC
Anti-ATP1B2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: ATP1B2 (ATPase, Na+/K+ transporting, beta 2 polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR