Anti-N-Myc (NCM II 100) Mouse Monoclonal Antibody [clone: NCM II 100]

Supplier: Merck Millipore (Oncogene)
OP13-100
OP13-100EA 746 GBP
OP13-100
Anti-N-Myc (NCM II 100) Mouse Monoclonal Antibody [clone: NCM II 100]
Antibodies
Type: Primary
Antigen: N-Myc (NCM II 100)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR