Anti-CAMP Mouse Monoclonal Antibody

Supplier: Genscript
A01509-100 A01509-200
GENSA01509-100EA 535 GBP
GENSA01509-100 GENSA01509-200
Anti-CAMP Mouse Monoclonal Antibody
Antibodies
Type: Tag
Antigen: CAMP (cathelicidin antimicrobial peptide)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa (Mouse)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR