Anti-TREX1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-29617 NBP2-29617SS
NOVUNBP2-29617EA 426 GBP
NOVUNBP2-29617 NOVUNBP2-29617SS
Anti-TREX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TREX1 (three prime repair exonuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR