Anti-MAS1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78444 NBP1-78444SS
NOVUNBP1-78444EA 642 GBP
NOVUNBP1-78444 NOVUNBP1-78444SS
Anti-MAS1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MAS1 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Primate.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: MAS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR