Anti-IGF1R Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77679 NBP1-77679SS
NOVUNBP1-77679EA 553 GBP
NOVUNBP1-77679 NOVUNBP1-77679SS
Anti-IGF1R Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal IGF1 Receptor Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Primate.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: IGF1R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR