Anti-HMOX1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77460 NBP1-77460SS
NOVUNBP1-77460EA 539 GBP
NOVUNBP1-77460 NOVUNBP1-77460SS
Anti-HMOX1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Heme Oxygenase 1 Antibody. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: HMOX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol HMOX1
Environmentally Preferable
Antigen name Heme Oxidase 1
Antigen synonyms Hsp32|heat shock protein 32|HMOX1|HO-1|heme orygenase 1
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 3162
Isotype
ImmunoFluorescence Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR