Anti-PKM Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-48308 NBP1-48308SS
NOVUNBP1-48308EA 621 GBP
NOVUNBP1-48308 NOVUNBP1-48308SS
Anti-PKM Rabbit Polyclonal Antibody
Antibodies
The PKM2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PKM2. This antibody reacts with human, mouse, rat. The PKM2 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: PKM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR