Anti-SLC31A2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-05199 NBP1-05199SS
NOVUNBP1-05199EA 539 GBP
NOVUNBP1-05199 NOVUNBP1-05199SS
Anti-SLC31A2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC31A2 (solute carrier family 31 (copper transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR